missing translation for 'onlineSavingsMsg'
Learn More

Medronic acid, MedChemExpress

Product Code. 16415146
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16415146 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16415146 Supplier Medchem Express Supplier No. HY108309
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Medronic acid (Methylenediphosphonic acid) is a methylene-substituted bisphosphonate. Medronic acid has affinity for and adheres to the surface of hydroxyapatite crystals in the bone matrix. Medronic acid accumulates at sites of osteoid mineralization and can be used, complexed with radioisotopes in bone imaging.

Medronic acid (Methylenediphosphonic acid) is a methylene-substituted bisphosphonate. Medronic acid has affinity for and adheres to the surface of hydroxyapatite crystals in the bone matrix. Medronic acid accumulates at sites of osteoid mineralization and can be used, complexed with radioisotopes in bone imaging.

Specifications

Chemical Name or Material Medronic acid
Physical Form Solid
CAS 1984-15-2
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula CH6O6P2
Synonym Methylenediphosphonic acid
Solubility Information DMSO : 250 mg/mL (1420.45 mM; Need ultrasonic)
SMILES O=P(O)(O)CP(O)(O)=O
Molecular Weight (g/mol) 176.0
Formula Weight 176.0
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.