missing translation for 'onlineSavingsMsg'
Learn More

Medroxyprogesterone acetate, MedChemExpress

Product Code. 16478326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478326 100 mg 100mg
16472146 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478326 Supplier Medchem Express Supplier No. HYB0469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Medroxyprogesterone acetate is a widely used synthetic steroid by its interaction with progesterone, androgen and glucocorticoid receptors.

Medroxyprogesterone acetate is a widely used synthetic steroid by its interaction with progesterone, androgen and glucocorticoid receptors.

Specifications

Chemical Name or Material Medroxyprogesterone acetate
Physical Form Powder
CAS 71-58-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C24H34O4
For Use With (Application) Cancer-programmed cell death
Synonym Medroxyprogesterone 17-acetate Farlutin
Solubility Information DMSO : 10 mg/mL (25.87 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.59 mM; ultrasonic and warming and heat to 80°C)
SMILES CC(O[C@@]([C@]12C)(CC[C@@]1([H])[C@@](C[C@H](C)C3=CC4=O)([H])[C@]([C@]3(CC4)C)([H])CC2)C(C)=O)=O
Molecular Weight (g/mol) 386.52
Formula Weight 386.52
Percent Purity 98.68%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.