Learn More
MEF2A Antibody (S06-9A6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19687-100ul
 View more versions of this product
Product Code. 18697733
  • 645.00 € / 100µL
Description
MEF2A Monoclonal antibody specifically detects MEF2A in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MEF2A
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S06-9A6
Unconjugated
ADCAD1, MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancerfactor 2A), MEF2, myocyte enhancer factor 2A, myocyte-specific enhancer factor 2A, RSRFC4, RSRFC9, Serum response factor-like protein 1
A synthetic peptide of human MEF2A (Uniprot # Q02078)
100 μg
Phospho Specific
4205
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.