missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEF2A/MEF2C Recombinant Rabbit Monoclonal Antibody (24GB1280)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557067
This item is not returnable.
View return policy
Description
MEF2A/MEF2C Recombinant Monoclonal Antibody for Western Blot, Flow
MEF2A is found in many muscle specific-genes mediating cellular functions in skeletal and cardiac muscle. MEF2C is a transcription activator which binds specifically to the MEF2 element present in the regulatory regions of many muscle-specific genes. Controls cardiac morphogenesis and myogenesis, and is also involved in vascular development. Plays an essential role in hippocampal-dependent learning and memory by suppressing the number of excitatory synapses and thus regulating basal and evoked synaptic transmission. Crucial for normal neuronal development, distribution, and electrical activity in the neocortex. Necessary for proper development of megakaryocytes and platelets and for bone marrow B lymphopoiesis. Required for B-cell survival and proliferation in response to BCR stimulation, efficient IgG1 antibody responses to T-cell-dependent antigens and for normal induction of germinal center B cells. May also be involved in neurogenesis and in the development of cortical architecture. Isoform 3 and isoform 4, which lack the repressor domain, are more active than isoform 1 and isoform 2.
Specifications
| MEF2A/MEF2C | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MEF2A | |
| ADCAD1; C5DELq14.3; DEL5q14.3; mef2; NEDHSIL; RSRFC4; RSRFC9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 309957, 4205, 4208, 499497 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1280 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q02078, Q06413, Q2MJT0 | |
| MEF2A, MEF2C | |
| A synthesized peptide derived from human MEF2A/MEF2C (aa 250-473). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction