missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEF2A Recombinant Rabbit Monoclonal Antibody (24GB85)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557090
This item is not returnable.
View return policy
Description
MEF2A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.
Specifications
| MEF2A | |
| Recombinant Monoclonal | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q02078, Q2MJT0, Q60929 | |
| MEF2A | |
| A synthesized peptide derived from human MEF2A (aa 465-507). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB85 | |
| Unconjugated | |
| MEF2A | |
| A430079H05Rik; ADCAD1; C5DELq14.3; DEL5q14.3; MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A); MADS box transcription enhancer factor 2, polypeptide C; mef2; Mef2a; MEF2C; myocyte enhancer factor 2a; myocyte enhancer factor 2C; Myocyte-specific enhancer factor 2A; myocyte-specific enhancer factor 2C; RSRFC4; RSRFC9; Serum response factor-like protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17258, 309957, 4205 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction