missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEG2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148615
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
MEG2 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal domain that shares a significant similarity with yeast SEC14, which is a protein that has phosphatidylinositol transfer activity and is required for protein secretion through the Golgi complex in yeast. This PTP was found to be activated by polyphosphoinositide, and is thought to be involved in signaling events regulating phagocytosis.
Specifications
| MEG2 | |
| Polyclonal | |
| Unconjugated | |
| PTPN9 | |
| Meg2; meg-2; protein tyrosine phosphatase, non-receptor type 9; protein-tyrosine phosphatase MEG2; Protein-tyrosine phosphatase, non-receptor type 9 (Protein-tyrosine phosphatase MEG2) (PTPase-MEG2); PTPase MEG2; PTPase-MEG2; PTPMEG2; PTPN9; Tyrosine-protein phosphatase non-receptor type 9 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 266611, 56294, 5780 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O35239, P43378, Q641Z2 | |
| PTPN9 | |
| E. coli-derived recombinant human PTP-MEG2, Met1-Gln593. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur