missing translation for 'onlineSavingsMsg'
Learn More

Megestrol acetate, MedChemExpress

Product Code. 16438477
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438477 5 g 5g
16401379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438477 Supplier Medchem Express Supplier No. HY13676
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Megestrol acetate is a synthetic and orally active progesteronal agent. Megestrol acetate is effective as an appetite stimulant for wasting syndromes such as cachexia. Megestrol acetate decreases nuclear and cytosol androgen receptors human BPH tissue. Megestrol acetate has the potential for HIV study and downregulates autophagic catabolic pathway.

Megestrol acetate is a synthetic and orally active progesteronal agent. Megestrol acetate is effective as an appetite stimulant for wasting syndromes such as cachexia. Megestrol acetate decreases nuclear and cytosol androgen receptors human BPH tissue. Megestrol acetate has the potential for HIV study and downregulates autophagic catabolic pathway.

Specifications

Chemical Name or Material Megestrol acetate
Physical Form Solid
CAS 595-33-5
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C24H32O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 20 mg/mL (52.01 mM; Need ultrasonic)
SMILES CC([C@@]1(OC(C)=O)CC[C@@]2([H])[C@]3([H])C=C(C)C4=CC(CC[C@]4(C)[C@@]3([H])CC[C@]12C)=O)=O
Molecular Weight (g/mol) 384.51
Formula Weight 384.51
Percent Purity 98.59%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.