missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEGF10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05646-20ul
This item is not returnable.
View return policy
Description
MEGF10 Polyclonal antibody specifically detects MEGF10 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MEGF10 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| KIAA1780, multiple EGF-like-domains 10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 26-160 of human MEGF10 (NP_115822.1). ADYTISGTLPHSNGGNANSHYFTNPSYHTLTQCATSPHVNNRDRMTVTKSKNNQLFVNLKNVNPGKRGPVGDCTGTLPADWKHGGYLNELGAFGLDRSYMGKSLKDLGKNSEYNSSNCSLSSSENPYATIKDPPV | |
| 20 μg | |
| Apoptosis, Cell Biology, Endocrinology, Stem Cells | |
| 84466 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction