Learn More
Invitrogen™ MEIS1 Recombinant Rabbit Monoclonal Antibody (24GB420)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557070
 View more versions of this product
Product Code. 30275995
  • 146.00 € / 20µL
Description
MEIS1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Homeobox genes, of which the most well-characterized category is represented by the HOX genes, play a crucial role in normal development. In addition, several homeoproteins are involved in neoplasia. This gene encodes a homeobox protein belonging to the TALE family of homeodomain-containing proteins.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MEIS1
Recombinant Monoclonal
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O00470, Q60954
MEIS1
A synthesized peptide derived from human MEIS1 (aa 320-390).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB420
Unconjugated
MEIS1
C530044H18Rik; cb224; etID37540.13; Evi8; Homeobox protein Meis1; homeobox protein Meis1b; id:ibd5078; leukemogenic homolog protein; Meis homeobox 1; Meis homeobox 1 b; Meis homeobox 1 S homeolog; meis1; Meis1, myeloid ecotropic viral integration site 1 homolog; meis1.1; meis1.S; MEIS1A; meis1b; MGC43380; myeloid ecotropic viral insertion site-1a protein; myeloid ecotropic viral integration 1; myeloid ecotropic viral integration site 1; transcription factor Meis1.1; wu:fb38b03; wu:fc02e11; wu:fd12a02; WUGSC:H_NH0444B04.1; XMeis1; XMeis1-1 protein; XMeis1-2 protein; XMeis1-4 protein
Rabbit
Affinity chromatography
RUO
17268, 4211, 686117
-20°C