missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS2 Antibody (PCRP-MEIS2-2B3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20520-100ug
This item is not returnable.
View return policy
Description
MEIS2 Monoclonal antibody specifically detects MEIS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| MEIS2 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2 | |
| Recombinant fragment of human MEIS2 | |
| 100 μg | |
| Vision | |
| 4212 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PCRP-MEIS2-2B3 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction