missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEKK4 Antibody (S05-8A5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19990-50ul
This item is not returnable.
View return policy
Description
MEKK4 Monoclonal antibody specifically detects MEKK4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MEKK4 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin | |
| EC 2.7.11.25, FLJ42439, MAPKKK4yeast, homolog of, MEKK 4, mitogen-activated protein kinase kinase kinase 4 | |
| A synthetic peptide of human MEKK4 (Uniprot # Q9Y6R4) | |
| 50 μg | |
| Angiogenesis, Cancer, Phospho Specific, Protein Kinase | |
| 4216 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S05-8A5 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction