missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melan-A/MART-1 Antibody (CL12864), Novus Biologicals™
Mouse Monoclonal Antibody
429.00 € - 611.00 €
Specifications
| Antigen | Melan-A/MART-1 |
|---|---|
| Clone | CL12864 |
| Dilution | Western Blot 1 μg/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 2-10 μg/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629814
|
Novus Biologicals
NBP3-18562-100ul |
100 μg |
611.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678024
|
Novus Biologicals
NBP3-18562-25ul |
25 μg |
429.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Melan-A/MART-1 Monoclonal antibody specifically detects Melan-A/MART-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Melan-A/MART-1 | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 2-10 μg/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A | |
| This antibody was developed against a recombinant protein of Q16655. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL12864 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Immunology | |
| PBS, pH 7.2, 40% glycerol | |
| 2315 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit