Learn More
Melan-A/MART-1 Antibody (CL12874) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18548UV
Product Code. 30303608
  • 770.00 € / 0.10mL
Description
Melan-A/MART-1 Monoclonal antibody specifically detects Melan-A/MART-1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Melan-A/MART-1
Monoclonal
DyLight 350
Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A
This antibody was developed using a recombinant protein derived from Q16655, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cellular Markers, Immunology
2315
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL12874
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified