missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melan-A/MART-1 Antibody (M2-7C10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
595.00 € - 643.00 €
Specifications
| Antigen | Melan-A/MART-1 |
|---|---|
| Clone | M2-7C10 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18487541
|
Novus Biologicals
NBP2-33148APC |
0.1 mL |
595.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18466401
|
Novus Biologicals
NBP2-33148APC-100ul |
100 μL |
643.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Melan-A/MART-1 Monoclonal antibody specifically detects Melan-A/MART-1 in Human, Equine, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| Melan-A/MART-1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Equine, Rat (Negative) | |
| Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A | |
| Recombinant Melan-A/MART-1 protein (Uniprot: Q16655) | |
| Primary | |
| Store at 4C in the dark. |
| M2-7C10 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| APC | |
| Mouse | |
| Cell Biology, Cellular Markers, Immunology | |
| PBS | |
| 2315 | |
| IgG2b λ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title