missing translation for 'onlineSavingsMsg'
Learn More

Melan-A/MART-1 Antibody (MLANA/1409R), Novus Biologicals™

Rabbit Monoclonal Antibody

353.00 € - 891.00 €

Specifications

Antigen Melan-A/MART-1
Clone MLANA/1409R
Concentration 0.2 mg/mL
Dilution Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18117709
View Documents
Novus Biologicals
NBP2-53301-20UG
20 μg
353.00 €
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18138259
View Documents
Novus Biologicals
NBP2-53301-100UG
100 μg
587.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18197699
View Documents
Novus Biologicals
NBP2-53301-200UG
200 μg
891.00 €
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Melan-A/MART-1 Monoclonal specifically detects Melan-A/MART-1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Melan-A/MART-1
0.2 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cytoskeleton Markers, Immunology
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
2315
Recombinant human full-length MLANA protein
Primary
Store at 4C.
MLANA/1409R
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Monoclonal
Purified
RUO
Human, Mouse, Rat
Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A
MLANA
IgG κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.