missing translation for 'onlineSavingsMsg'
Learn More

Melanin, MP Biomedicals™

Product Code. 11497622 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497622 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11497622 Supplier MP Biomedicals Supplier No. 0215534380
  • ONLINE EXCLUSIVE PRICE
    117.00 €
    110.25 € / 100mg
    Save 6.75 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 8049-97-6
Molecular Formula C18H10N2O4
Molecular Weight (g/mol) 318.288
InChI Key XUMBMVFBXHLACL-UHFFFAOYSA-N
Synonym melanin, phaeomelanins, d0wq4g, melanin from sepia officinalis, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1; 2,5 .0; 13,17 .0; 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1^ 2,5 .0^ 13,17 .0^ 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone
PubChem CID 6325610
SMILES CC1=C2C3=C(C4=CNC5=C(C(=O)C(=O)C(=C45)C3=CN2)C)C(=O)C1=O

Specifications

Color Brown to Black
Quantity 100 mg
Formula Weight 1780
Physical Form Granular Powder
Chemical Name or Material Melanin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.