missing translation for 'onlineSavingsMsg'
Learn More

Melatonin, MedChemExpress

Product Code. 16480768
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480768 5 g 5g
16492039 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16480768 Supplier Medchem Express Supplier No. HYB0075
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. Afficher la politique du retour.

MedChemExpress Melatonin is a hormone made by the pineal gland that can activates melatonin receptor. Melatonin plays a role in sleep and possesses important antioxidative and anti-inflammatory properties. Melatonin is a novel selective ATF-6 inhibitor and induces human hepatoma cell apoptosis through COX-2 downregulation. Melatonin attenuates palmitic acid-induced (HY-N0830) mouse granulosa cells apoptosis via endoplasmic reticulum stress.

Melatonin is a hormone made by the pineal gland that can activates melatonin receptor. Melatonin plays a role in sleep and possesses important antioxidative and anti-inflammatory properties. Melatonin is a novel selective ATF-6 inhibitor and induces human hepatoma cell apoptosis through COX-2 downregulation. Melatonin attenuates palmitic acid-induced (HY-N0830) mouse granulosa cells apoptosis via endoplasmic reticulum stress.

Spécification

Chemical Name or Material Melatonin
CAS 73-31-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C13H16N2O2
Quantity 5 g
Synonym N-Acetyl-5-methoxytryptamine
Solubility Information DMSO : 100 mg/mL (430.51 mM; Need ultrasonic) ∣Ethanol : 12.5 mg/mL (53.81 mM; Need ultrasonic) ∣H2O : 1.1 mg/mL (4.74 mM; Need ultrasonic)
SMILES CC(NCCC1=CNC2=C1C=C(OC)C=C2)=O
Molecular Weight (g/mol) 232.28
Formula Weight 232.28
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Afficher plus Afficher moins

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.