missing translation for 'onlineSavingsMsg'
Learn More

Melatonin, MedChemExpress

Product Code. 16492039
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492039 1 g 1g
16480768 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492039 Supplier Medchem Express Supplier No. HYB0075
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Melatonin is a hormone made by the pineal gland that can activates melatonin receptor. Melatonin plays a role in sleep and possesses important antioxidative and anti-inflammatory properties. Melatonin is a novel selective ATF-6 inhibitor and induces human hepatoma cell apoptosis through COX-2 downregulation. Melatonin attenuates palmitic acid-induced (HY-N0830) mouse granulosa cells apoptosis via endoplasmic reticulum stress.

Melatonin is a hormone made by the pineal gland that can activates melatonin receptor. Melatonin plays a role in sleep and possesses important antioxidative and anti-inflammatory properties. Melatonin is a novel selective ATF-6 inhibitor and induces human hepatoma cell apoptosis through COX-2 downregulation. Melatonin attenuates palmitic acid-induced (HY-N0830) mouse granulosa cells apoptosis via endoplasmic reticulum stress.

Specifications

Chemical Name or Material Melatonin
CAS 73-31-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C13H16N2O2
Quantity 1 g
Synonym N-Acetyl-5-methoxytryptamine
Solubility Information DMSO : 100 mg/mL (430.51 mM; Need ultrasonic) ∣Ethanol : 12.5 mg/mL (53.81 mM; Need ultrasonic) ∣H2O : 1.1 mg/mL (4.74 mM; Need ultrasonic)
SMILES CC(NCCC1=CNC2=C1C=C(OC)C=C2)=O
Molecular Weight (g/mol) 232.28
Formula Weight 232.28
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.