missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MELK Recombinant Rabbit Monoclonal Antibody (24GB3530)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557071
This item is not returnable.
View return policy
Description
MELK Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MELK is a serine/threonione kinase which is related to the AMPK family that has been linked to the cell cycle. MELK has also been shown to be overexpressed in cancer and has been identified has a potential target for cancer therapeutics. MELK has a broad substrate specificity; phosphorylates BCL2L14, CDC25B, MAP3K5/ASK1 and ZNF622. Acts as an activator of apoptosis by phosphorylating and activating MAP3K5/ASK1. Acts as a regulator of cell cycle, notably by mediating phosphorylation of CDC25B, promoting localization of CDC25B to the centrosome and the spindle poles during mitosis. Plays a key role in cell proliferation and carcinogenesis. Required for proliferation of embryonic and postnatal multipotent neural progenitors. Phosphorylates and inhibits BCL2L14, possibly leading to affect mammary carcinogenesis by mediating inhibition of the pro-apoptotic function of BCL2L14. Also involved in the inhibition of spliceosome assembly during mitosis by phosphorylating ZNF622, thereby contributing to its redirection to the nucleus. May also play a role in primitive hematopoiesis.
Specifications
| MELK | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MELK | |
| AI327312; hMELK; HPK38; Kiaa0175; Maternal embryonic leucine zipper kinase; Melk; mKIAA0175; MPK38; OTTHUMP00000046113; pEg3 kinase; Pk38; Protein kinase Eg3; protein kinase PK38; RP11-8N6.1; Tyrosine-protein kinase MELK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17279, 362510, 9833 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3530 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14680, Q61846 | |
| MELK | |
| A synthesized peptide derived from human MELK (aa 1-36). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction