missing translation for 'onlineSavingsMsg'
Learn More

Meloxicam, MedChemExpress

Product Code. 16468196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468196 100 mg 100mg
16442076 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468196 Supplier Medchem Express Supplier No. HYB0261
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Meloxicam is a non-steroidal antiinflammatory agent, inhibits COX activity, with IC50s of 0.49 μM and 36.6 μM for COX-2 and COX-1, respectively.

Meloxicam is a non-steroidal antiinflammatory agent, inhibits COX activity, with IC50s of 0.49 μM and 36.6 μM for COX-2 and COX-1, respectively.

Specifications

Chemical Name or Material Meloxicam
CAS 71125-38-7
Color Yellowish Green
Purity Grade Notes Research
Molecular Formula C14H13N3O4S2
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (142.29 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(O)C2=CC=CC=C2S(N1C)(=O)=O)NC3=NC=C(C)S3
Molecular Weight (g/mol) 351.4
Formula Weight 351.4
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.