missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM (10X), no glutamine

Product Code. 11570566 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
5000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11570566 500 mL 500mL
11580566 10 x 500 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11570566 Supplier Gibco™ Supplier No. 21430020
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MEM (10X), no glutamine

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow’s MEM, MEM α, DMEM, and Temin’s Modification. MEM is available with Earle’s salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle’s salts.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Concentrated forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 X
Form Liquid
Product Type MEM Medium
With Additives Phenol Red
Without Additives No Glutamine, No HEPES, No Sodium Bicarbonate
For Use With (Application) Mammalian cell culture
Quantity 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 1.2 to 1.8
Regulatory Status In-vitro Diagnostic Use
Content And Storage 2 to 8°C
Description MEM Medium, 500mL, 10X Concentration, Liquid Form, With Phenol Red, 1.2 to 1.8 pH, 12 Months Shelf Life, 2400 to 3000mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Soluble in Water, Used for Mammalian Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.