missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM α, GlutaMAX™ Supplement, no nucleosides

Product Code. 11524466 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
500mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packungsgröße
11524466 500 mL 500mL
11534466 10 x 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11524466 Lieferant Gibco™ Lieferanten-Nr. 32561029
Follow the Workflow - Explore the Savings
Use promo code 27967
Alle Werbeaktionen anzeigen
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

MEM α, GlutaMAX™ Supplement, no nucleosides

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. Gibco™ MEM α with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. This product is made with Earle’s salts.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ MEM α is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ MEM α product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Spezifikation

Concentration 1 X
Form Liquid
Product Type Minimum Essential Medium α±
With Additives GlutaMAX, Phenol Red, Sodium Pyruvate
Without Additives No Ribonucleosides, No Deoxyribonucleosides
For Use With (Application) Mammalian cell culture
Quantity 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 7.0 to 7.4
Content And Storage 2 to 8°C
Description Minimum Essential Medium α±, 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With Glutamax™, Phenol Red, Sodium Pyruvate, 12 Months Shelf Life, 280 to 320mOsm/kg Osmolality, For Mammalian Cell Culture
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.