missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ MEM, Hanks' Balanced Salts

Description
Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
Dual-Site cGMP Manufacturing
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). This MEM formulation does not require a CO2 environment to maintain physiological pH.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
Dual-Site cGMP Manufacturing
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). This MEM formulation does not require a CO2 environment to maintain physiological pH.
Specifications
Specifications
| Form | Liquid |
| Product Type | MEM Hanks' Balanced Salts |
| With Additives | Hanks’ Balanced Salt solution (HBSS), Glutamine, Phenol Red |
| Without Additives | No HEPES |
| For Use With (Application) | Mammalian cell culture |
| Quantity | 10 x 500 mL |
| Certifications/Compliance | ISO 13485 standards, FDA |
| Regulatory Status | In-vitro Diagnostic Use |
| Content And Storage | 2 to 8°C |
| Description | MEM Hanks' Balanced Salts, Liquid Form, With Hanks' salts, L-glutamine, Phenol Red, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?