missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM, HEPES, GlutaMAX™ Supplement
GREENER_CHOICE

Product Code. p-7184674
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 12569089

Brand: Gibco™ 42360032

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Requires

Supplementation

Minimum Essential Medium is commonly used of all cell culture media

MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. Gibco MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification.

  • Available with Earle's salts for use in CO2 incubator, or with Hank's salts for use without CO2
  • Gibco MEM with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
  • Modified as follows: with GlutaMAX™, HEPES, phenol red
  • Made with Earle's salts
  • Contains no proteins, lipids, or growth factors
  • Requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • Uses sodium bicarbonate buffer system (2.2g/L) and therefore requires 5-10% CO2 environment to maintain physiological pH
  • Sustainable packaging

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Cell Type Primary Rat Astrocytes
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type MEM (Minimum Essential Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives Low Glucose, GlutaMAX, HEPES, Phenol Red
Without Additives No Sodium Pyruvate
Product Line Gibco, GlutaMAX
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.