missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM, NEAA, no glutamine
GREENER_CHOICE

Product Code. 11583397
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
500mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11583397 500 mL 500mL
11593397 10 x 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11583397 Supplier Gibco™ Supplier No. 10370047
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

MEM, NEAA, no glutamine

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification. MEM is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle's salts.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

Dual-Site cGMP Manufacturing
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco™ MEM product made in our Scotland facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10%Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Minimum Essential Medium NEAA
With Additives Non Essential Amino Acids (NEAA), Phenol Red
Without Additives No Glutamine, No HEPES
For Use With (Application) For cell culture, mammalian cell culture, In-vitro cultivation
Quantity 500 mL
Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 2 to 8°C
Description Minimum Essential Medium (MEM), NEAA, 500mL, Liquid Form, Standard Serum Supplementation, 2 to 8°C Storage Temperature, 12 Months Shelf Life, Without L-glutamine, HEPES, Certified to ISO 13485 Standards, For In-vitro Diagnostic Use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.