missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM α, no nucleosides, powder

Product Code. 10442312
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 10442312

Brand: Gibco™ 12000014

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

MEM α, no nucleosides, powder

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. This product is made with Earle’s salts.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ MEM α is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM α product made in our Grand Island facility (12000-022). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type MEM α± Medium
With Additives Glutamine, Phenol Red, Sodium Pyruvate
Without Additives No Sodium Bicarbonate
For Use With (Application) Mammalian cell culture
Quantity 1 L
Certifications/Compliance ISO 13485 standards, FDA
pH 3.2 to 3.9
Regulatory Status For in-vitro diagnostic use
Content And Storage 2 to 8°C
Description MEM α± Medium, 1L, Powdered Form, With L-glutamine, Phenol Red, Sodium Pyruvate, 3.2 to 3.9 pH, 36 Months Shelf Life, 240 to 280mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.