missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM, powder

Product Code. 10378292 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10378292 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10378292 Supplier Gibco™ Supplier No. 61100053
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MEM, powder

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow’s MEM, MEM α, DMEM, and Temin’s Modification. MEM is available with Earle’s salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle’s salts.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM product made in our Grand Island facility (61100-061). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Powder forms of Gibco™ L-15 cell culture medium require pH adjustment and filtration at the time of preparation (see protocol for details).
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Minimum Essential Medium
With Additives Glutamine
Without Additives No HEPES, No Phenol Red, No Sodium Bicarbonate, No Sodium Pyruvate
For Use With (Application) Mammalian cell culture
Quantity 1 L
Certifications/Compliance ISO 13485 standards, FDA
pH 5.6 to 6.0
Content And Storage 2 to 8°C
Description Minimum Essential Medium, 1L, Powder, 5.6 to 6.0 pH, With L-Glutamine, Phenol Red, 36 Months Shelf Life, 240 to 280mOsm/kg Osmolality, Standard Serum Supplementation, For Mammalian Cell Culture

For In Vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.