missing translation for 'onlineSavingsMsg'
Learn More

Menadione, 98.76%, MP Biomedicals™

Product Code. 11453490 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453490 25 g 25g
11463490 100 g 100g
11473490 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453490 Supplier MP Biomedicals Supplier No. 0210225925
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 58-27-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
InChI Key MJVAVZPDRWSRRC-UHFFFAOYSA-N
Synonym menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
PubChem CID 4055
ChEBI CHEBI:28869
IUPAC Name 2-methylnaphthalene-1,4-dione
SMILES CC1=CC(=O)C2=CC=CC=C2C1=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 172.2
Percent Purity 98.76%
Physical Form Powder
Chemical Name or Material Menadione
compliance-icons
Product Identifier
  • Menadione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.