missing translation for 'onlineSavingsMsg'
Learn More

Menaquinone 9, TRC

Product Code. 30008996
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008996 10 mg 10mg
30012756 5 mg 5mg
30011093 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008996 Supplier Toronto Research Chemicals Supplier No. TRCM21861010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 523-39-7
Molecular Formula C55 H78 O2
Molecular Weight (g/mol) 771.21
Synonym 2-Methyl-3-[(2E,6E,10E,14E,18E,22E,26E,30E)-3,7,11,15,19,23,27,31,35-nonamethyl-2,6,10,14,18,22,26,30,34-hexatriacontanonaen-1-yl]-1,4-naphthalenedione, Vitamin K2(45) (6CI), MK9, Menaquinone MK 9, Vitamin MK 9;
IUPAC Name 2-methyl-3-[(2E,6E,10E,14E,18E,21E,25E,29E)-3,7,11,15,19,22,26,30,34-nonamethylpentatriaconta-2,6,10,14,18,21,25,29,33-nonaenyl]naphthalene-1,4-dione
SMILES CC(=CCC\C(=C\CC\C(=C\CC\C(=C\C\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CC1=C(C)C(=O)c2ccccc2C1=O)\C)\C)\C)\C)\C)\C)\C)\C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C55H78O2/c1-41(2)21-14-22-42(3)23-15-24-44(5)29-18-32-47(8)37-38-48(9)33-19-30-45(6)27-16-25-43(4)26-17-28-46(7)31-20-34-49(10)39-40-51-50(11)54(56)52-35-12-13-36-53(52)55(51)57/h12-13,21,23,26-27,29,31,33,35-37,39H,14-20,22,24-25,28,30,32,34,38,40H2,1-11H3/b42-23+,43-26+,44-29+,45-27+,46-31+,47-37+,48-33+,49-39+
Formula Weight 770.6002
Percent Purity >95%
Chemical Name or Material Menaquinone 9
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.