missing translation for 'onlineSavingsMsg'
Learn More

Menthol 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30313810
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313810 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313810 Supplier Dr. Ehrenstorfer Supplier No. DREA14866000AL1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Menthol
CAS 1490-04-6
Analyte or Component Names Menthol
Shipping Condition Room Temperature
Molecular Formula C10 H20 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Cyclohexanol, 5-methyl-2-(1-methylethyl)-, Menthol (8CI), 5-Methyl-2-(1-methylethyl)cyclohexanol, 1-Methyl-4-isopropyl-3-cyclohexanol, 2-(2-Propyl)-5-methyl-1-cyclohexanol, 2-Isopropyl-5-methylcyclohexan-1-ol, 2-Isopropyl-5-methylcyclohexanol, 3-Hydroxy-p-menthane, 5-Methyl-2-isopropylcyclohexanol, Menthyl alcohol, p-Menthan-3-ol
InChI Formula InChI=1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3
SMILES CC(C)C1CCC(C)CC1O
IUPAC Name 5-methyl-2-propan-2-ylcyclohexan-1-ol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 156.27
Quantity 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.