missing translation for 'onlineSavingsMsg'
Learn More

Menthoxypropane-1,2-diol, TRC

Product Code. 30133081
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133081 1 g 1g
30007662 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133081 Supplier Toronto Research Chemicals Supplier No. TRCM8850351G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87061-04-9
Molecular Formula C13H26O3
Molecular Weight (g/mol) 230.3
Synonym TK 10, 3-[[5-Methyl-2-(1-methylethyl)cyclohexyl]oxy]-1,2-propanediol
IUPAC Name 3-(5-methyl-2-propan-2-ylcyclohexyl)oxypropane-1,2-diol
SMILES CC(C)C1CCC(C)CC1OCC(O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H26O3/c1-9(2)12-5-4-10(3)6-13(12)16-8-11(15)7-14/h9-15H,4-8H2,1-3H3
Formula Weight 230.1882
Chemical Name or Material Menthoxypropane-1,2-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.