missing translation for 'onlineSavingsMsg'
Learn More

Meperfluthrin, Dr. Ehrenstorfer

Product Code. 30318862
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318862 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318862 Supplier Dr. Ehrenstorfer Supplier No. DREC14868000
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Meperfluthrin
CAS 915288-13-0
Analyte or Component Names Meperfluthrin
Shipping Condition Room Temperature
Molecular Formula C17 H16 Cl2 F4 O3
Certifications/Compliance ISO 17034
Synonym Meperfluthrin, [2,3,5,6-Tetrafluoro-4-(methoxymethyl)phenyl]methyl (1R,3S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate, 2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl (1R,3S)-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, 2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl (1R)-trans-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate
InChI Formula InChI=1S/C17H16Cl2F4O3/c1-17(2)9(4-10(18)19)11(17)16(24)26-6-8-14(22)12(20)7(5-25-3)13(21)15(8)23/h4,9,11H,5-6H2,1-3H3/t9-,11+/m1/s1
SMILES COCc1c(F)c(F)c(COC(=O)[C@@H]2[C@@H](C=C(Cl)Cl)C2(C)C)c(F)c1F
IUPAC Name [2,3,5,6-tetrafluoro-4-(methoxymethyl)phenyl]methyl (1R,3S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Molecular Weight (g/mol) 415.21
Quantity 25 mg
Formula Weight 414.0413 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.