Learn More
Mercury Organometallic Standards, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep ORG-HG8-2Z
Code : 04
Additional Details : CAS Number : 7439-97-6 Weight : 0.09000kg
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Hg | 1000μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Fluid | |
7439-97-6,64742-65-0 | |
99.9 | |
MFCD00011035 | |
[Hg] | |
200.59 | |
CHEBI:16170 | |
Base Oil 75 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Mercury Oil Standard | |
99.9 | |
Hg | |
QSHDDOUJBYECFT-UHFFFAOYSA-N | |
mercury | |
23931 | |
Triple-leached LDPE bottle | |
50 g | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
Safety and Handling
- 1000 ug/g Mercury in Base Oil 75
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : petroleum distilates, mercury
Product intended for laboratory use only