missing translation for 'onlineSavingsMsg'
Learn More

Mercury Standard For ICP, 10μg/mL in 5% HNO3, SPEX CertiPrep™

Product Code. 11989532 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11989532 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11989532 Supplier SPEX Certiprep Supplier No. PLHG21AY
  • ONLINE EXCLUSIVE PRICE
    63.90 €
    56.39 € / 125mL
    Save 7.51 €
    11% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Hg10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7439-97-6, 7697-37-2, 7732-18-5
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Molecular Formula Hg
MDL Number MFCD00011035
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [Hg]
IUPAC Name mercury
Solvent or Matrix 5% HNO3
Molecular Weight (g/mol) 200.59
PubChem CID 23931
Quantity 125 mL
ChEBI CHEBI:16170
Grade Assurance
Chemical Name or Material Mercury Standard
Show More Show Less
compliance-icons
Product Identifier
  • 10 µg/mL Mercury
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.