missing translation for 'onlineSavingsMsg'
Learn More

Melford Meropenem Trihydrate

Product Code. 30018489
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018489 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018489 Supplier Melford Supplier No. M625001.0
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Meropenem is a broad-spectrum antibiotic.

Meropenem is a broad-spectrum antibiotic. It is a carbapenem antibiotic that inhibits bacterial cell wall synthesis. It acts by binding to penicillin-binding proteins (PBPs), enzymes involved in the final stages of peptidoglycan synthesis in the bacterial cell wall. This interference leads to structural instability and eventual cell lysis.

Meropenem may be used as a tool to study antibiotic susceptibility patterns of bacteria or as a selective agent in culture media to isolate bacteria sensitive to this antibiotic.

Specifications

Chemical Name or Material Meropenem Trihydrate
CAS 119478-56-7
Quantity 1 g
Molecular Formula C17H25N3O5S-3H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.