missing translation for 'onlineSavingsMsg'
Learn More

Meropenem trihydrate, 98-101%

Product Code. 15437809
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437809 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15437809 Supplier Thermo Scientific Alfa Aesar Supplier No. J66446.MF
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 119478-56-7 | C17H25N3O5S | 383.46 g/mol

Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.

Solubility
Soluble in aqueous solution at approximately 5mg/ml

Notes
TRUSTED_SUSTAINABILITY

Specifications

Type Meropenem Trihydrate
Physical Form Powder
CAS 119478-56-7
Color White to Yellow
Quantity 50 mg
Flash Point >110°C (230°F)
Molecular Formula C17H25N3O5S
MDL Number MFCD08600005,MFCD00864966
Boiling Point 627°C
Merck Index 14,5900
Solubility Information Soluble in aqueous solution at approximately 5mg/mL
InChI Key FSTGLKRHSQANLP-PQTSNVLCSA-N
SMILES C[C@@H](O)[C@@H]1[C@H]2[C@@H](C)C(S[C@H]3CC[C@H](N3)C(=O)N(C)C)=C(N2C1=O)C(O)=O
IUPAC Name (4R,5S,6S)-3-{[(2S,5S)-5-(dimethylcarbamoyl)pyrrolidin-2-yl]sulfanyl}-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid
Molecular Weight (g/mol) 383.46
PubChem CID 441130
ChEBI CHEBI:43968
Formula Weight 383.46
Percent Purity 98 to 101%
Chemical Name or Material Meropenem trihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Meropenem trihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Meropenem trihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.