missing translation for 'onlineSavingsMsg'
Learn More

MES (Fine White Crystals), Fisher BioReagents

Product Code. 10419123
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10419123 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10419123 Supplier Fisher BioReagents Supplier No. BP300100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Commonly used buffering agent

This biological buffer has a usable pH range of 5.5 to 6.7.
TRUSTED_SUSTAINABILITY

Specifications

CAS 145224-94-8
Melting Point 300°C
Color White
pH 3.5 to 4.1
Physical Form Powder or Solid
Quantity 100 g
DNase DNase free
Assay Percent Range ≥98 %
Identification Pass Test
Molecular Formula C6H15NO5S
Linear Formula OCH2CH2NCH2CH2(CH2CH2SO3H)
Packaging Glass Bottle
Merck Index 15, 5972
Vapor Pressure Negligible
Synonym 2-(4-Morpholino)ethane Sulfonic Acid, 2-(N-Morpholino)ethanesulfonic acid hydrate
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
SMILES C1COCCN1CCS(=O)(=O)O.O
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
Molecular Weight (g/mol) 213.248
PubChem CID 16218417
Formula Weight 213.2
Percent Purity ≥98%
Chemical Name or Material MES
Grade DNase- and RNase-Free
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.