missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES hydrate, 99+%, for biochemistry

Product Code. 10206230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10206230 25 g Plastic bottle 25g
10052190 100 g Plastic bottle 100g
10266130 500 g Plastic bottle 500g
10721414 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10206230 Supplier Thermo Scientific Chemicals Supplier No. 172590250
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1266615-59-1 | C6H13NO4S | 195.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-59-1
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.23
MDL Number MFCD00149409
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 4-Morpholinoethanesulfonic acid hydrate, MES
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid
SMILES OS(=O)(=O)CCN1CCOCC1

Specifications

Absorbance (0.1M in H2O) 1 cm cell, (0.1M in H2O) 1 cm cell at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
Absorbance Conditions (0.1M aq. soln.) (1 cm cell)
Melting Point 243.0°C
Color White
pH 2.5 to 4.0 (1%, H2O, 25°C)
Physical Form Needle-like Crystals or Powder
Quantity 25 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Loss on Drying 9.5% max. (105°C, 3 hrs)
Packaging Plastic bottle
Merck Index 14, 5902
Solubility Information Solubility in water: soluble.
Formula Weight 195.23
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material MES hydrate, 0.99
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.