missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MESDC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | MESDC2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-4 uL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661586
|
Novus Biologicals
NBP2-99668-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661616
|
Novus Biologicals
NBP2-99668-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MESDC2 Polyclonal antibody specifically detects MESDC2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| MESDC2 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 23184 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-4 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BOCA, KIAA0081Renal carcinoma antigen NY-REN-61, MESDLDLR chaperone MESD, mesoderm development candidate 2Mesoderm development protein | |
| Produced in rabbits immunized with purified, recombinant Human MESDC2 (Accession#: NP_055969.1; Ala 34-Lys 230) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title