missing translation for 'onlineSavingsMsg'
Learn More

Mesitylenesulfonic acid dihydrate, 98+%

Product Code. 11456996
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456996 25 g 25g
11466996 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456996 Supplier Thermo Scientific Alfa Aesar Supplier No. B25321.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 835617-36-2 | C9H16O5S | 236.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 835617-36-2
Molecular Formula C9H16O5S
Molecular Weight (g/mol) 236.282
MDL Number MFCD00149535
InChI Key LIKWMDGAWXCECN-UHFFFAOYSA-N
Synonym mesitylenesulfonic acid dihydrate, 2-mesitylenesulfonic acid dihydrate, 2,4,6-trimethylbenzenesulfonic acid dihydrate, benzenesulfonic acid, 2,4,6-trimethyl-, dihydrate, mesitylenesulfonicaciddihydrate, mesitylenesulfonic acid dihydrate, 98+%, benzenesulfonic acid,2,4,6-trimethyl-, hydrate 1:2, 2,4,6-trimethylbenzenesulfonic acid, oxamethane, oxamethane
PubChem CID 15620344
IUPAC Name 2,4,6-trimethylbenzenesulfonic acid;dihydrate
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)O)C.O.O

Specifications

Melting Point 74°C to 77°C
Quantity 25 g
UN Number UN2585
Beilstein 1961501
Formula Weight 236.29 (200.26 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Mesitylenesulfonic acid dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.