missing translation for 'onlineSavingsMsg'
Learn More

meso-Erythritol, MedChemExpress

Product Code. 16454376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16454376 Supplier Medchem Express Supplier No. HY100551
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress meso-Erythritol is a sugar alcohol that occurs naturally in a variety of foods (e.g., pear, watermelon), is 60-80% as sweet as sucrose, and is an approved low-calorie sweetener food additive.

meso-Erythritol is a sugar alcohol that occurs naturally in a variety of foods (e.g., pear, watermelon), is 60-80% as sweet as sucrose, and is an approved low-calorie sweetener food additive.

Specifications

Chemical Name or Material meso-Erythritol
Physical Form Solid
CAS 149-32-6
Color White to Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C4H10O4
Solubility Information DMSO : 100 mg/mL (818.87 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (818.87 mM)
SMILES OC[C@@H](O)[C@@H](O)CO
Molecular Weight (g/mol) 122.12
Formula Weight 122.12
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.