missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesosulfuron, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Mesosulfuron |
| CAS | 400852-66-6 |
| Analyte or Component Names | Mesosulfuron |
| Shipping Condition | +20°C |
| Molecular Formula | C16 H19 N5 O9 S2 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 2-[[[[(4,6-Dimethoxy-2-pyrimidinyl)amino]carbonyl]amino]sulfonyl]-4-[[(methylsulfonyl)amino]methyl]benzoic acid, AE-F 154851 |
| InChI Formula | InChI=1S/C16H19N5O9S2/c1-29-12-7-13(30-2)19-15(18-12)20-16(24)21-32(27,28)11-6-9(8-17-31(3,25)26)4-5-10(11)14(22)23/h4-7,17H,8H2,1-3H3,(H,22,23)(H2,18,19,20,21,24) |
| SMILES | COc1cc(OC)nc(NC(=O)NS(=O)(=O)c2cc(CNS(=O)(=O)C)ccc2C(=O)O)n1 |
| IUPAC Name | 2-[(4,6-dimethoxypyrimidin-2-yl)carbamoylsulfamoyl]-4-(methanesulfonamidomethyl)benzoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?