Learn More
Mesothelin Antibody (CL13956) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24589R
Product Code. 30301635
  • 737.00 € / 0.10mL
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mesothelin
Monoclonal
DyLight 550
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein
This antibody was developed using a recombinant protein derived from Q13421, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
10232
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL13956
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified