missing translation for 'onlineSavingsMsg'
Learn More

Mesothelin (Mesothelial Marker) Mouse, Clone: MSLN/2131, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 10232-MSM1-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959707

  • 485.00 € / 100µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Mesothelin (Mesothelial Marker)
Cell Surface and Secreted
MSLN/2131
Unconjugated
Mouse
40kDa (Mature form); 69kDa (Precursor)
Protein A/G purified from Bioreactor concentrate.
RUO
10232
IgG2b κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant fragment (apprx. aa 273-407) of human Mesothelin (MSLN) protein
CAPAN-2 or HeLa cells. Lung Mesothelioma.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers