missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Trichloroethane in Methanol 10μg/mL, Fisher Chemical™

Artikelnummer. 12960284 Shop alle Fisher Chemical produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
10 mL
Pakningsstørrelse:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
12960284 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 12960284 Leverandør Fisher Chemical Leverandørnr. P855860

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Tekniske data

Packaging Ampoule
CAS 79-00-5
Physical Form Liquid
Molecular Formula C2H3Cl3
Concentration 10 μg/mL
MDL Number MFCD00000852
UN Number 1230
InChI Key UBOXGVDOUJQMTN-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES ClCC(Cl)Cl
IUPAC Name 1,1,2-trichloroethane
Solvent or Matrix Methanol
Molecular Weight (g/mol) 133.40
PubChem CID 6574
Quantity 10 mL
ChEBI CHEBI:36018
Chemical Name or Material 1,1,2-Trichloroethane
Vis mere Vis mindre
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Standard solution 1,1,2-Trichloroethane 10ug/ml in methanol
Signalord
  • Danger
Farekategori
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Faresætning
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.