missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trichloropropane in Methyl-tert.butylether 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12910314 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12910314 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12910314 Supplier Fisher Chemical Supplier No. P856310
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 96-18-4
Physical Form Liquid
Molecular Formula C3H5Cl3
Concentration 100 μg/mL
MDL Number MFCD00000946
InChI Key CFXQEHVMCRXUSD-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES ClCC(Cl)CCl
IUPAC Name 1,2,3-trichloropropane
Solvent or Matrix MTBE
Molecular Weight (g/mol) 147.42
PubChem CID 7285
Quantity 1 mL
ChEBI CHEBI:34036
Chemical Name or Material 1,2,3-Trichloropropane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.