missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane D4 in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12925384 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12925384 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12925384 Supplier Fisher Chemical Supplier No. P818640
  • ONLINE EXCLUSIVE PRICE
    139.89 €
    118.40 € / 1mL
    Save 21.49 €
    15% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 17060-07-0
Molecular Formula C2H4Cl2
Concentration 100 μg/mL
InChI Key WSLDOOZREJYCGB-LNLMKGTHSA-N
SMILES C(CCl)Cl
IUPAC Name 1,2-dichloro-1,1,2,2-tetradeuterioethane
Solvent or Matrix Methanol
Molecular Weight (g/mol) 102.978
PubChem CID 12197860
Quantity 1 mL
Chemical Name or Material 1,2-Dichloroethane D4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.