missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tekniske data
Tekniske data
| Packaging | Ampoule(s) |
| CAS | 141-93-5 |
| Molecular Formula | C10H14 |
| Concentration | 100 μg/mL |
| UN Number | 1230 |
| InChI Key | AFZZYIJIWUTJFO-UHFFFAOYSA-N |
| SMILES | CCC1=CC(=CC=C1)CC |
| IUPAC Name | 1,3-diethylbenzene |
| Solvent or Matrix | Methanol |
| Molecular Weight (g/mol) | 134.222 |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- Standard solution 1,3-Diethylbenzene 100ug/ml in methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?