missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metabolite 1-Chloro-2-fluorobenzene in Methanol 100μg/mL, Fisher Chemical™
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Packaging | Ampoule(s) |
| CAS | 348-51-6 |
| Molecular Formula | C6H4ClF |
| Concentration | 100 μg/mL |
| MDL Number | MFCD00000533 |
| InChI Key | ZCJAYDKWZAWMPR-UHFFFAOYSA-N |
| SMILES | FC1=CC=CC=C1Cl |
| IUPAC Name | 1-chloro-2-fluorobenzene |
| Solvent or Matrix | Methanol |
| Molecular Weight (g/mol) | 130.55 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?