missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tribromophenol in Ethanol 1000μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 118-79-6 |
| Molecular Formula | C6H3Br3O |
| Concentration | 1000 μg/mL |
| MDL Number | MFCD00002150 |
| InChI Key | BSWWXRFVMJHFBN-UHFFFAOYSA-N |
| Type | Organic Single-component Standard Solution |
| SMILES | OC1=C(Br)C=C(Br)C=C1Br |
| IUPAC Name | 2,4,6-tribromophenol |
| Solvent or Matrix | Ethanol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction